Learn More
AdipoGen Rifamycin AF
SDP
Supplier:  AdipoGen AGCN20321G005
Encompass_Preferred
Chemical. CAS 13292-22-3. Formula C38H47NO13. MW 725.8. Semisynthetic. Ansamycin antibiotic. Intermediate of rifampicin. Selective inhibitor of bacterial DNA-dependent RNA polymerase RNAP. Effective against mycobacteria, and are therefore used in research of tuberculosis, leprosy and mycobacterium avium complex MAC infections.
Catalog No. 50-256-2600