Learn More
AdipoGen Rifamycin AF-EPTAPI
SDP
Supplier:  AdipoGen AGCN20326M010
Encompass_Preferred
Chemical. CAS 41971-22-6. Formula C49H70N4O12. MW 907.1. Semisynthetic. Ansamycin antibiotic. Selective inhibitor of bacterial DNA-dependent RNA polymerase RNAP. Effective against mycobacteria, and are therefore used in research of tuberculosis, leprosy and mycobacterium avium complex MAC infections.
Catalog No. 50-256-2590