Learn More
AdipoGen Rifamycin AF-K28259
SDP
Supplier:  AdipoGen AGCN20324M050
Encompass_Preferred
Chemical. CAS 51707-27-8. Formula C45H54N4O12S. MW 875. Semisynthetic. Ansamycin antibiotic. Hydrazone of 3-formylrifamycin SV. Selective inhibitor of bacterial DNA-dependent RNA polymerase RNAP.
Catalog No. 50-256-2593