Learn More
AdipoGen Rifamycin AF-K43033
SDP
Supplier:  AdipoGen AGCN20320M010
Encompass_Preferred
Chemical. CAS 57369-86-5. Formula C41H55N3O13. MW 797.9. Semisynthetic. Ansamycin antibiotic. Hydrazone of 3-formylrifamycin SV. Selective inhibitor of bacterial DNA-dependent RNA polymerase RNAP.
Catalog No. 50-256-2603