Learn More
AdipoGen Rifamycin AF-K55517
SDP
Supplier:  AdipoGen AGCN20322M050
Encompass_Preferred
Chemical. CAS 51707-26-7. Formula C42H54N4O12S. MW 839. Semisynthetic. Ansamycin antibiotic. Hydrazone of 3-formylrifamycin SV. Selective inhibitor of bacterial DNA-dependent RNA polymerase RNAP.
Catalog No. 50-256-2597