Learn More
AdipoGen Rifamycin AF-K91725
SDP
Catalog No. 502562588
Encompass_Preferred
Chemical. CAS 13292-38-1. Formula C45H55N3O12. MW 829.9. Semisynthetic. Ansamycin antibiotic. Selective inhibitor of bacterial DNA-dependent RNA polymerase RNAP. Effective against mycobacteria, and are therefore used in research of tuberculosis, leprosy and mycobacterium avium complex MAC infections.
Catalog No. 50-256-2588 Supplier AdipoGen Supplier No. AGCN20327M010
Add to Cart
Add to Cart
Chemical. CAS 13292-38-1. Formula C45H55N3O12. MW 829.9. Semisynthetic. Ansamycin antibiotic. Selective inhibitor of bacterial DNA-dependent RNA polymerase RNAP. Effective against mycobacteria, and are therefore used in research of tuberculosis, leprosy and mycobacterium avium complex MAC infections.