Learn More
AdipoGen Rifamycin AF-O13
SDP
Supplier:  AdipoGen AGCN20336M050
Encompass_Preferred
Chemical. CAS 35225-13-9. Formula C46H64N2O13. MW 853. Semisynthetic. Ansamycin antibiotic. Potent inhibitor of nucleic acid polymerizing enzymes and of some hormone receptors. Selective inhibitor of bacterial DNA-dependent RNA polymerase RNAP. Inhibitor of RNA-dependent DNA polymerase of MSV. Effective against mycobacteria, and therefore might be used in research of tuberculosis, leprosy and mycobacterium avium complex MAC infections.
Catalog No. 50-256-2570