Learn More
AdipoGen Rifamycin AF-pNFI
SDP
Supplier:  AdipoGen AGCN20338M050
Encompass_Preferred
Chemical. CAS 39976-91-5. Formula C44H52N4O14. MW 860.9. Semisynthetic. Ansamycin antibiotic. Hydrazone of 3-formylrifamycin SV. Selective inhibitor of bacterial DNA-dependent RNA polymerase RNAP.
Catalog No. 50-256-2566