Learn More
AdipoGen Rifamycin AG
SDP
Catalog No. 502562584
Encompass_Preferred
Chemical. CAS 38128-51-7. Formula C38H49N5O11. MW 751.8. Semisynthetic. Ansamycin antibiotic. Selective inhibitor of bacterial DNA-dependent RNA polymerase RNAP. Effective against mycobacteria, and are therefore used in research of tuberculosis, leprosy and mycobacterium avium complex MAC infections.
Catalog No. 50-256-2584 Supplier AdipoGen Supplier No. AGCN20329M050
Chemical. CAS 38128-51-7. Formula C38H49N5O11. MW 751.8. Semisynthetic. Ansamycin antibiotic. Selective inhibitor of bacterial DNA-dependent RNA polymerase RNAP. Effective against mycobacteria, and are therefore used in research of tuberculosis, leprosy and mycobacterium avium complex MAC infections.