Learn More
AdipoGen Rifamycin AMI-DA
SDP
Catalog No. 502562582
Encompass_Preferred
Chemical. CAS 17096-02-5. Formula C40H53N3O12. MW 767.9. Semisynthetic. Ansamycin antibiotic. Selective inhibitor of bacterial DNA-dependent RNA polymerase RNAP. Effective against mycobacteria, and are therefore used in research of tuberculosis, leprosy and mycobacterium avium complex MAC infections.
Catalog No. 50-256-2582 Supplier AdipoGen Supplier No. AGCN20330M050
Add to Cart
Add to Cart
Chemical. CAS 17096-02-5. Formula C40H53N3O12. MW 767.9. Semisynthetic. Ansamycin antibiotic. Selective inhibitor of bacterial DNA-dependent RNA polymerase RNAP. Effective against mycobacteria, and are therefore used in research of tuberculosis, leprosy and mycobacterium avium complex MAC infections.