Learn More
AdipoGen Rifamycin AMP-DA
SDP
Supplier:  AdipoGen AGCN20331M050
Encompass_Preferred
Chemical. CAS 16783-99-6. Formula C41H56N4O11. MW 780.9. Semisynthetic. Ansamycin antibiotic. Selective inhibitor of bacterial DNA-dependent RNA polymerase RNAP. Effective against mycobacteria, and are therefore used in research of tuberculosis, leprosy and mycobacterium avium complex MAC infections.
Catalog No. 50-256-2580