Learn More
AdipoGen Rifamycin O
SDP
Supplier:  AdipoGen AGCN20333M500
Encompass_Preferred
Chemical. CAS 14487-05-9. Formula C39H47NO14. MW 753.8. Semisynthetic. Ansamycin antibiotic. Selective inhibitor of bacterial DNA-dependent RNA polymerase RNAP. Effective against mycobacteria, and are therefore used in research of tuberculosis, leprosy and mycobacterium avium complex MAC infections.
Catalog No. 50-256-2576