Learn More
AdipoGen Rifamycin PR-14
SDP
Supplier:  AdipoGen AGCN20334M050
Encompass_Preferred
Chemical. CAS 21240-38-0. Formula C43H54N2O12. MW 790.9. Semisynthetic. Ansamycin antibiotic. Selective inhibitor of bacterial DNA-dependent RNA polymerase RNAP. Effective against mycobacteria, and are therefore used in research of tuberculosis, leprosy and mycobacterium avium complex MAC infections.
Catalog No. 50-256-2574