Learn More
AdipoGen Rifamycin PR-3
SDP
Supplier:  AdipoGen AGCN20335M010
Encompass_Preferred
Chemical. CAS 21288-98-2. Formula C43H54N2O13. MW 806.9. Semisynthetic. Ansamycin antibiotic. Selective inhibitor of bacterial DNA-dependent RNA polymerase RNAP. Effective against mycobacteria, and are therefore used in research of tuberculosis, leprosy and mycobacterium avium complex MAC infections.
Catalog No. 50-256-2573