Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Rifamycin S | 13553-79-2 | MFCD06198807 | 98.8% | 695.75 | C37H45NO12 | 25G
SDP

Catalog No. 5000237271
Encompass_Preferred

Rifamycin S is an ansamycin-class antibiotic (CAS 13553-79-2) commonly used as a reference compound in microbiology and biochemical research. It inhibits bacterial RNA polymerase and shows activity against various Gram-positive organisms, including MRSA and Mycobacterium species. The material is provided as a high-purity solid suitable for analytical, microbiological, and biochemical assays.

  • High purity: 98.8%
  • Chemical formula: C37H45NO12; molecular weight: 695.75 g/mol
  • Intended use: research reagent and antibiotic reference compound
  • Available formats: solid packs and a 10 mM solution in DMSO
  • Safety and documentation: SDS and technical data available

Catalog No. 50-002-37271 Supplier Medchemexpress LLC Supplier No. HY12536525G
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Rifamycin S is an ansamycin-class antibiotic (CAS 13553-79-2) commonly used as a reference compound in microbiology and biochemical research. It inhibits bacterial RNA polymerase and shows activity against various Gram-positive organisms, including MRSA and Mycobacterium species. The material is provided as a high-purity solid suitable for analytical, microbiological, and biochemical assays.

  • High purity: 98.8%
  • Chemical formula: C37H45NO12; molecular weight: 695.75 g/mol
  • Intended use: research reagent and antibiotic reference compound
  • Available formats: solid packs and a 10 mM solution in DMSO
  • Safety and documentation: SDS and technical data available

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.