Learn More
Medchemexpress LLC Rifamycin S | 13553-79-2 | MFCD06198807 | 98.8% | 695.75 g·mol⁻¹ | C37H45NO12 | 100G
SDP
Catalog No. 5000235693
Encompass_Preferred
Rifamycin S is an antibiotic compound of the rifamycin family (CAS 13553-79-2) supplied as a solid for research and analytical use. It is an oxidized rifamycin active against Gram-positive bacteria and commonly used as a reference standard and in antibiotic activity assays. Molecular formula C37H45NO12, molecular weight 695.75 g·mol⁻¹.
  • High purity (98.8%) suitable for analytical and assay use.
  • Characterized chemical data for reproducible results.
  • Suitable for microbiology and antibiotic activity studies.
  • Multiple pack sizes available to meet small-scale or bulk requirements.
  • Safety and handling information provided in the SDS.
Catalog No. 50-002-35693 Supplier Medchemexpress LLC Supplier No. HY125365100G
Rifamycin S is an antibiotic compound of the rifamycin family (CAS 13553-79-2) supplied as a solid for research and analytical use. It is an oxidized rifamycin active against Gram-positive bacteria and commonly used as a reference standard and in antibiotic activity assays. Molecular formula C37H45NO12, molecular weight 695.75 g·mol⁻¹.
  • High purity (98.8%) suitable for analytical and assay use.
  • Characterized chemical data for reproducible results.
  • Suitable for microbiology and antibiotic activity studies.
  • Multiple pack sizes available to meet small-scale or bulk requirements.
  • Safety and handling information provided in the SDS.