Learn More
Medchemexpress LLC Rifamycin S | 13553-79-2 | MFCD06198807 | 98.8% | 695.75 g/mol | C37H45NO12 | 250MG
SDP
Catalog No. 5000235986
Encompass_Preferred
Rifamycin S (CAS 13553-79-2) is a quinone antibiotic from the rifamycin family used as a research reagent. It exhibits activity against Gram-positive bacteria and is reported to generate reactive oxygen species and inhibit microsomal lipid peroxidation. The compound is supplied in solid form and as a DMSO solution for laboratory and analytical use.
Catalog No. 50-002-35986 Supplier Medchemexpress LLC Supplier No. HY125365250MG
Rifamycin S (CAS 13553-79-2) is a quinone antibiotic from the rifamycin family used as a research reagent. It exhibits activity against Gram-positive bacteria and is reported to generate reactive oxygen species and inhibit microsomal lipid peroxidation. The compound is supplied in solid form and as a DMSO solution for laboratory and analytical use.