Learn More
Medchemexpress LLC Rifamycin S | 13553-79-2 | MFCD06198807 | 98.8% | 695.8 g/mol | C37H45NO12 | 50G
SDP
Catalog No. 5000237399
Encompass_Preferred
Rifamycin S is a rifamycin-class antibiotic research compound that inhibits bacterial DNA-dependent RNA polymerase. It is supplied as a purified reference material for biochemical and microbiological studies, and is used in antimicrobial research, mechanism-of-action experiments, and assay development.
  • High purity suitable for research experiments (98.8%).
  • Acts as an RNA polymerase inhibitor for bacterial studies.
  • Available in multiple pack sizes for small-scale and bulk use.
  • Offered as solid and as DMSO solution options for convenience.
  • Covers essential identifiers: CAS 13553-79-2, formula C37H45NO12, molecular weight 695.8 g/mol.
  • Datasheet, COA, and SDS are available to support use and safety compliance.
Catalog No. 50-002-37399 Supplier Medchemexpress LLC Supplier No. HY12536550G
Rifamycin S is a rifamycin-class antibiotic research compound that inhibits bacterial DNA-dependent RNA polymerase. It is supplied as a purified reference material for biochemical and microbiological studies, and is used in antimicrobial research, mechanism-of-action experiments, and assay development.
  • High purity suitable for research experiments (98.8%).
  • Acts as an RNA polymerase inhibitor for bacterial studies.
  • Available in multiple pack sizes for small-scale and bulk use.
  • Offered as solid and as DMSO solution options for convenience.
  • Covers essential identifiers: CAS 13553-79-2, formula C37H45NO12, molecular weight 695.8 g/mol.
  • Datasheet, COA, and SDS are available to support use and safety compliance.