Learn More
AdipoGen Rifamycin S 8-Methyl-
SDP
Supplier:  AdipoGen AGCN20337M050
Encompass_Preferred
Chemical. CAS 42898-13-5. Formula C38H47NO12. MW 709.8. Semisynthetic. Ansamycin antibiotic. Selective inhibitor of bacterial DNA-dependent RNA polymerase RNAP. Effective against mycobacteria, and are therefore used in research of tuberculosis, leprosy and mycobacterium avium complex MAC infections.
Catalog No. 50-256-2568