Learn More
Medchemexpress LLC HY-B1907 50mg Medchemexpress, Rifamycin (sodium) CAS:14897-39-3 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HYB190750MG
Encompass_Preferred
Medchemexpress, HY-B1907 50mg Rifamycin (sodium) CAS:14897-39-3 Rifamycin sodium (Rifamycin SV monosodium) belongs to the family of ansamycin antibiotics and has been isolated from the fermentation of A. mediterranei or its mutants. Rifamycin sodium displays a broad spectrum of antibiotic activity against Gram-positive and, to a lesser extent, Gram-negative bacteria. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-197-0172