Learn More
Cayman Chemical Rifamycin SV (sodium salt hydrate), Purity: ≥98%, 500 mg
SDP
Catalog No. 502994285
Encompass_Preferred
A broad-spectrum antibiotic that inhibits bacterial DNA-dependent RNA polymerase
Catalog No. 50-299-4285 Supplier Cayman Chemical Supplier No. 21441500 MG
A broad-spectrum antibiotic that inhibits bacterial DNA-dependent RNA polymerase