Learn More
Cayman Chemical Rifapentine, Purity: ≥98%, CAS: 61379-65-5, 100 mg
SDP
Catalog No. 502982432
Encompass_Preferred
A long-lasting rifamycin antibiotic that is effective against Mycobacterium species, including M. tuberculosis as well as experimental infections caused by S. aureus, S. pyogenes group A, S. pneumoniae, and K. pneumoniae
Catalog No. 50-298-2432 Supplier Cayman Chemical Supplier No. 20307100 MG
A long-lasting rifamycin antibiotic that is effective against Mycobacterium species, including M. tuberculosis as well as experimental infections caused by S. aureus, S. pyogenes group A, S. pneumoniae, and K. pneumoniae