Learn More
Apexbio Technology LLC Rifapentine, 61379-65-5, MFCD00866806, 25 mg
SDP
Catalog No. 501011532
Encompass_Preferred
MF: C47H64N4O12, MW: 877.03, Solubility: 30.25mg/mL in DMSO. Rifapentine is an antibiotic drug used in the treatment of tuberculosis.
Catalog No. 50-101-1532 Supplier Apexbio Technology LLC Supplier No. B212725
MF: C47H64N4O12, MW: 877.03, Solubility: 30.25mg/mL in DMSO. Rifapentine is an antibiotic drug used in the treatment of tuberculosis.