Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thermo Scientific Chemicals Rifapentine, 95%

Catalog No. AC461900250
Change view
Click to view available options
Quantity:
25 mg
100 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AC461900250 25 mg
AC461901000 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC461900250 Supplier Thermo Scientific Chemicals Supplier No. 461900250
Add to Cart

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 61379-65-5
Molecular Formula C47H64N4O12
Molecular Weight (g/mol) 877.03
InChI Key RISDZDVHYCURHD-QLWTZYDESA-N
PubChem CID 131846483

Specifications

Melting Point 179°C to 180°C
Assay Percent Range 94% min. (HPLC)
Quantity 25 mg
Formula Weight 877.03
Percent Purity 95%
Chemical Name or Material Rifapentine
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
EINECSNumber 262-743-9

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.