Learn More
Medchemexpress LLC Rifapentine (Standard) | 61379-65-5 | The compound is the grade of analytical standard, which is the reference standard supplied assay. It is commonly used in qualitative, quantitative and methodological research experiments in HPLC, GC and MS. | 877.03 | 10 MG
SDP
Catalog No. 5000365137
Encompass_Preferred
Rifapentine (Standard) serves as the analytical standard for Rifapentine. This product is designed for research and analytical applications, and is recognized as an antibiotic utilized in tuberculosis research.
  • Analytical standard for Rifapentine
  • Intended for research and analytical applications
  • Antibiotic used in tuberculosis research
  • Reference standard supplied assay
  • Suitable for qualitative, quantitative and methodological research in HPLC, GC, and MS
Catalog No. 50-003-65137 Supplier Medchemexpress LLC Supplier No. 10MGHYB0269R
Rifapentine (Standard) serves as the analytical standard for Rifapentine. This product is designed for research and analytical applications, and is recognized as an antibiotic utilized in tuberculosis research.
  • Analytical standard for Rifapentine
  • Intended for research and analytical applications
  • Antibiotic used in tuberculosis research
  • Reference standard supplied assay
  • Suitable for qualitative, quantitative and methodological research in HPLC, GC, and MS