Learn More
Apexbio Technology LLC Rifaximin (Xifaxan), 500mg. Cas: 80621-81-4 MFCD: MFCD00864973. RNA synthesis inhibitor, PXR activator
SDP
Catalog No. 501905155
Encompass_Preferred
Rifaximin, a rifamycin derivative, is a nonabsorbable antibiotic. Rifaximin inhibits RNA synthesis by binding the subunit of the bacterial DNA-dependent RNA polymerase. Other sizes are available. Please inqury us for quote.
Catalog No. 50-190-5155 Supplier Apexbio Technology LLC Supplier No. A8512500
Rifaximin, a rifamycin derivative, is a nonabsorbable antibiotic. Rifaximin inhibits RNA synthesis by binding the subunit of the bacterial DNA-dependent RNA polymerase. Other sizes are available. Please inqury us for quote.