Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Rift Valley Fever Virus Polymerase Synthetic Peptide

Catalog No. PEP1612 Shop All Thermo Scientific Products
Change view
Click to view available options
Quantity:
50 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PEP1612 50 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PEP1612 Supplier Invitrogen™ Supplier No. PEP1612
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Synthetic Peptide

This peptide corresponds to 16 amino acids near the amino terminus of Rift Valley Fever Virus Polymerase. For use as a blocking peptide in Western blot, incubate the peptide with equal volume of antibody for 30 min at 37°C.

Rift Valley Fever (RFV) virus is an arthropod-borne virus endemic to Africa that infects humans and animals that is transmitted predominantly by mosquitoes. During human infections, symptoms can range from benign fever to severe encephalitis and fatal hepatitis with hemorrhagic fever. The Bunyaviridae family of viruses to which the RVF virus belongs are spherical enveloped viruses with a tripartite RNA genome of negative or ambisense polarity. The three segments are referred to as the L, M, and S segments. The L and M segments are negative polarity and code fore the L-dependent RNA polymerase and glycoprotein precursor respectively. The S segment is of ambisense polarity and encodes the nucleoprotein and non-structural proteins.
TRUSTED_SUSTAINABILITY

Specifications

Species Virus
Purification Method purified
Common Name Rift Valley Fever Virus Polymerase
Concentration 200 μg/mL
Content And Storage -20°C
Description Synthetic Peptide
Format Liquid
Formulation PBS with 0.1% BSA and 0.02% sodium azide; pH 7.2
For Use With (Application) Blocking Assay, Control
Gene Alias RVV; RVV Polymerase
Lyophilized No
Preservative Sodium azide
Product Type Peptide
Quantity 50 μg
Regulatory Status RUO
Storage Buffer PBS
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.