Learn More
Invitrogen™ Rift Valley Fever Virus Synthetic Peptide
Description
This peptide corresponds to 19 amino acids near the carboxy terminus of RVF virus.
Specifications
Specifications
| Species | Virus |
| Purification Method | purified |
| Common Name | Rift Valley Fever Virus |
| Concentration | 200 μg/mL |
| Content And Storage | -20°C |
| Description | Synthetic Peptide |
| Format | Liquid |
| Formulation | PBS with 0.1% BSA and 0.02% sodium azide; pH 7.2 |
| For Use With (Application) | Blocking Assay, Control |
| Gene Alias | RVV |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.