Learn More
Cayman Chemical RIG-I helIcase domaInhumn50ug
SDP
Catalog No. 502934304
Encompass_Preferred
Source: Recombinant N-terminal histidine-tagged RIG-I helicase domain purified from E. coli • Amino acids: 232-794 • MW: 66.47 kDa
Catalog No. 50-293-4304 Supplier Cayman Chemical Supplier No. 2562050 UG
Source: Recombinant N-terminal histidine-tagged RIG-I helicase domain purified from E. coli • Amino acids: 232-794 • MW: 66.47 kDa