Learn More
Enzo Life Sciences RIG-I (human) polyclonal antibody (AT111) (100µg)
SDP
Supplier:  Enzo Life Sciences ALX210932C100
Encompass_Preferred
Host: Rabbit. Immunogen: Recombinant human RIG-I (aa 201-713). Source: Purified from rabbit serum. Species reactivity: Human. Applications: IP, WB. Purity: Protein A-affinity purified. Formulation: Liquid. In PBS containing 0.02% sodium azide. Handling: Avoid freeze/thaw cycles. After opening, prepare aliquots and store at -20°C.
Catalog No. 50-200-8188