Learn More
Cell Signaling Technology Rig-I Pathway Antibody Sampler Kit, 1 Kit
SDP
Catalog No. 502730858
Encompass
The Rig-I Pathway Antibody Sampler Kit offers a cost-effective solution for analyzing the activation status and total protein levels of key members within the Rig-I signaling pathway. This comprehensive kit contains a selection of primary antibodies, each provided in a volume sufficient for two western blot experiments.
  • Includes primary antibodies for Rig-I, MDA-5, MAVS, IRF-3, TBK1/NAK, and IKKε
  • Supports multiple applications including western blotting, immunofluorescence, flow cytometry, and immunoprecipitation
  • Includes HRP-linked secondary antibody for convenient detection
  • Supplied in storage buffer with 50% glycerol and less than 0.02% sodium azide
Catalog No. 50-273-0858 Supplier Cell Signaling Technology Supplier No. 8348T
The Rig-I Pathway Antibody Sampler Kit offers a cost-effective solution for analyzing the activation status and total protein levels of key members within the Rig-I signaling pathway. This comprehensive kit contains a selection of primary antibodies, each provided in a volume sufficient for two western blot experiments.
  • Includes primary antibodies for Rig-I, MDA-5, MAVS, IRF-3, TBK1/NAK, and IKKε
  • Supports multiple applications including western blotting, immunofluorescence, flow cytometry, and immunoprecipitation
  • Includes HRP-linked secondary antibody for convenient detection
  • Supplied in storage buffer with 50% glycerol and less than 0.02% sodium azide