Learn More
Medchemexpress LLC Rig012 | 2642218-43-5 | ≥99.0% | C23H21NO3 | 10MG
SDP
Catalog No. 5000258876
Encompass_Preferred
RIG012 is a small-molecule antagonist of the RIG-I innate immune receptor. It inhibits RIG-I ATPase activity (IC50 = 0.71 μM, NADH-coupled ATPase assay) and suppresses downstream signaling, including interferon-β and ISG expression. Supplied as a research-grade solid for in vitro studies; not for human or diagnostic use.
  • Potent RIG-I inhibition (IC50 0.71 μM).
  • Activity measured by NADH-coupled ATPase assay.
  • Suppresses IFN-β and ISG expression in cellular assays.
  • Supplied as a purified small-molecule for research use.
  • Product datasheet and safety data sheet available for handling information.
  • Useful for studying innate immune signaling and antiviral responses.
Catalog No. 50-002-58876 Supplier Medchemexpress LLC Supplier No. HY14712410MG
RIG012 is a small-molecule antagonist of the RIG-I innate immune receptor. It inhibits RIG-I ATPase activity (IC50 = 0.71 μM, NADH-coupled ATPase assay) and suppresses downstream signaling, including interferon-β and ISG expression. Supplied as a research-grade solid for in vitro studies; not for human or diagnostic use.
  • Potent RIG-I inhibition (IC50 0.71 μM).
  • Activity measured by NADH-coupled ATPase assay.
  • Suppresses IFN-β and ISG expression in cellular assays.
  • Supplied as a purified small-molecule for research use.
  • Product datasheet and safety data sheet available for handling information.
  • Useful for studying innate immune signaling and antiviral responses.