Learn More
BINDER™ Right Access Port (50 mm)
SDP
Catalog No. 08111536 Shop All BINDER Inc. Products
Change view
Click to view available options
Material:
Steel
Catalog No. Material
08-111-536 Steel
1 options
Catalog No. 08-111-536 Supplier BINDER™ Supplier No. 80121373
Add to Cart
Add to Cart
For Use With: FED 400
  • For use on right
  • Includes silicone plug
  • For use with a wide range of BINDER™ incubators and chambers
Specifications
Description Access port with silicone plug - 50 mm, right
For Use With (Equipment) BINDER Chambers FED 400
Material Steel
Type Access Port
Includes Silicone plug
Diameter (Metric) Inside 50 mm