Learn More
Glass Expansion Thermo 6000/7000 Series Duo Right Angle Ball Joint Adaptor
SDP
Catalog No. 502703178
Encompass_Preferred
Glass right angle ball joint adaptor compatible with Thermo 6000/7000 Duo Series ICP-OES.Glass Expansion is ISO 9001:2015 certified by AB Certification, No. AU4501-9001
Catalog No. 50-270-3178 Supplier Glass Expansion Supplier No. 218092761
Glass right angle ball joint adaptor compatible with Thermo 6000/7000 Duo Series ICP-OES.Glass Expansion is ISO 9001:2015 certified by AB Certification, No. AU4501-9001