Learn More
Access Technologies 22 gauge right angle infusion set 3/4 in., Pack of 12
Catalog No. NC9666688
Encompass
22 gauge right angle infusion set 3/4 in. needle with 12 in. extension. Our infusion sets have a right angle "Huber" Point Needle and grip block bonded to Tygon Tubing with a female luer lock. These infusion sets are supplied individually sterile and packaged per dozen. We can custom make our sets to securely fit your swivels and any other application that might be needed.
Catalog No. NC9666688 Supplier Access Technologies Supplier No. RA227512
22 gauge right angle infusion set 3/4 in. needle with 12 in. extension. Our infusion sets have a right angle "Huber" Point Needle and grip block bonded to Tygon Tubing with a female luer lock. These infusion sets are supplied individually sterile and packaged per dozen. We can custom make our sets to securely fit your swivels and any other application that might be needed.