Learn More
Specifications
Specifications
| Format | Insert |
| Product Type | Booklet |
| Includes | (25) Training Booklets |
| Width (Metric) | 172.72 mm |
| Quantity | Pack of 25 Each |
| Material | Paper |
| Color | Black/Red/Yellow on Blue |
| Languages | Spanish |
| For Use With (Application) | Hazard Communication |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.