Learn More
Waters Corp Waters Guard Connector, Rigid Connector Assembly, 0.020, HPLC
Catalog No. NC0706067 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
The Guard Connector is a rigid connector assembly designed for connecting guard columns to analytical and preparative HPLC columns. It ensures a secure, leak-free connection, minimizes dead volume, and maintains chromatographic efficiency. This connector helps protect the analytical column from particulate matter and contaminants, extending its lifespan and ensuring reliable separations. It is suitable for use with 19 mm ID, 20 mm ID, and larger prep columns.
  • Rigid connector assembly
  • Size 0.020
  • Compatible with 19 mm ID, 20 mm ID, and larger prep columns
  • Suitable for use in HPLC, LC/MS, and GC systems
Catalog No. NC0706067 Supplier Waters Corp Supplier No. 405000981
The Guard Connector is a rigid connector assembly designed for connecting guard columns to analytical and preparative HPLC columns. It ensures a secure, leak-free connection, minimizes dead volume, and maintains chromatographic efficiency. This connector helps protect the analytical column from particulate matter and contaminants, extending its lifespan and ensuring reliable separations. It is suitable for use with 19 mm ID, 20 mm ID, and larger prep columns.
  • Rigid connector assembly
  • Size 0.020
  • Compatible with 19 mm ID, 20 mm ID, and larger prep columns
  • Suitable for use in HPLC, LC/MS, and GC systems