Learn More
MSC Value Collection Pulley Block, Malleable Iron, 2 in. Sheave, 1/2 in. Fibrous Rope, 1 ea
Catalog No. 5000512190
Encompass
This rigid eye single sheave pulley is designed for lifting and pulling applications using fibrous rope. Constructed with a durable malleable iron body and a galvanized finish, this pulley block provides reliable utility for various rigging tasks.
  • Features a 2 in. outside diameter sheave
  • Compatible with 1/2 in. diameter fibrous rope
  • Engineered with a fixed eye attachment type
  • Equipped with a sleeve bearing for smooth operation
  • Includes one groove for rope guidance
Catalog No. 50-005-12190 Supplier MSC Supplier No. 67328765
This rigid eye single sheave pulley is designed for lifting and pulling applications using fibrous rope. Constructed with a durable malleable iron body and a galvanized finish, this pulley block provides reliable utility for various rigging tasks.
  • Features a 2 in. outside diameter sheave
  • Compatible with 1/2 in. diameter fibrous rope
  • Engineered with a fixed eye attachment type
  • Equipped with a sleeve bearing for smooth operation
  • Includes one groove for rope guidance