Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Fisherbrand™ Rigidwall Cleanroom

Catalog No. 15050989 Shop All Fisherbrand Products
Change view
Click to view available options
Length (English):
8 ft.
12 ft.
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Length (English)
15-050-989 8 ft.
15-050-991 12 ft.
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 15-050-989

Please work with Sales Representative for a quote.

Rigidwall cleanrooms combine class and functionality offering an elegant solution for most any cleanroom application. Easy to construct. Easy to clean.

  • Easy to construct, secure, and easy to clean
  • With wall panels providing full visibility from floor to ceiling, these rooms provide a comfortable working environment.
  • The modular nature of this system provides opportunity for expansion as your operation grows and offers the ability to relocate the cleanroom altogether should needs shift within your organization.
  • Our Rigidwall cleanrooms meet ISO 4 to ISO 8 air cleanliness ratings.

Specifications

Product Type Cleanroom
Filter Type 19 HEPA
Autoclavable Not Autoclavable
Height (English) 12 ft.
Length (English) 8 ft.
Width (English) 8 ft.
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.