Learn More
Axygen™ RIGIPLATE™ 96-Well PCR Microplate, Full Skirt
Catalog No. 14224602 Shop All Axygen Products
Change view
Click to view available options
Color:
Clear
Catalog No. Color
14-224-602 Clear
1 options
Catalog No. 14-224-602 Supplier Axygen™ Supplier No. PCR96EPFCC
Get Up to $2,500 in Products with Purchase
View all promotions
Add to Cart
Add to Cart
Standard ANSI/SLAS microplate footprint available in 96-and 384-well formats.
The RIGIPLATE PCR Microplates have an industry standard footprint enabling the compatibility with automated liquid handlers and most thermal cyclers. Each microplate is engineered to stay warp-free, provide precise positioning for automation, and deliver pressure resistance without distortion.
Features
  • Standard ANSI/SLAS microplate footprint available in 96-and 384-well formats
  • Polycarbonate frame that reduces plate distortion at both high and low temperatures
  • Thin-walled polypropylene wells allowing rapid and efficient heat transfer during PCR
  • Raised rim design to facilitate secure sealing
  • Manufactured in an ISO 13485 and ISO 8 clean room facility
Benefits
  • Improved thermal cycling: The combination of materials enhances thermal performance, resulting in more reliable PCR results
  • Easy sample handling: Sturdy construction prevents distortion and shrinkage and enables efficient sample handling
  • Consistent results: Well-to-well consistency ensures accurate data across experiments
TRUSTED_SUSTAINABILITY
Specifications
Barcode No Barcode
Certifications/Compliance ANSI/SLAS Footprint
Format Standard
Array 8 x 12
Color Clear
Product Type PCR Microplate
Skirt Style Full Skirt
Material Polycarbonate, Polypropylene
Volume (Metric) Well 200 μL
No. of Wells 96
Sterility Non-sterile
Surface Treatment Non-coated
Well Shape Conical Bottom (V-well)