Learn More
Medchemexpress LLC HY-12037A 5mg Medchemexpress, Rigosertib CAS:592542-59-1 Purity:>98%
SDP
Catalog No. 502028841
Encompass_Preferred
Medchemexpress, HY-12037A 5mg Rigosertib CAS:592542-59-1 Rigosertib (ON-01910) is a multi-kinase inhibitor and a selective anti-cancer agent, which induces apoptosis by inhibition the PI3 kinase/Akt pathway, promots the phosphorylation of histone H2AX and induces G2/M arrest in cell cycle[2]. Rigosertib is a selective and non-ATP-competitive inhibitor of PLK1 with an IC50 of 9 nM[3]. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-202-8841 Supplier Medchemexpress LLC Supplier No. HY12037A5MG
Medchemexpress, HY-12037A 5mg Rigosertib CAS:592542-59-1 Rigosertib (ON-01910) is a multi-kinase inhibitor and a selective anti-cancer agent, which induces apoptosis by inhibition the PI3 kinase/Akt pathway, promots the phosphorylation of histone H2AX and induces G2/M arrest in cell cycle[2]. Rigosertib is a selective and non-ATP-competitive inhibitor of PLK1 with an IC50 of 9 nM[3]. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.