Learn More
Apexbio Technology LLC Rigosertib sodium salt, 1225497-78-8, MFCD11655911, 5 mg
SDP
Catalog No. 501013663
Encompass_Preferred
MF: C21H24NNaO8S, MW: 473.47, Synonyms: ON-01910, Estybon. Solubility: 23.65mg/mL in DMSO. Rigosertib (ON-01910,Estybon) is a potent, specific PLK1 inhibitor with IC50 value of 9nM.
Catalog No. 50-101-3663 Supplier Apexbio Technology LLC Supplier No. A14045
MF: C21H24NNaO8S, MW: 473.47, Synonyms: ON-01910, Estybon. Solubility: 23.65mg/mL in DMSO. Rigosertib (ON-01910,Estybon) is a potent, specific PLK1 inhibitor with IC50 value of 9nM.