Learn More
Medchemexpress LLC Rigosertib sodium (ON-01910 sodium) | 592542-60-4 | 99.6% | 50 MG
SDP
Catalog No. 5000231894
Encompass_Preferred
Rigosertib sodium is a research reagent and multi-kinase inhibitor used in preclinical studies to investigate cell cycle regulation, DNA damage response, and apoptosis. It is provided as a solid powder for preparation of stock solutions and is supplied in milligram-scale packs for laboratory use.
  • Multi-kinase inhibitor targeting PLK and PI3K pathways
  • Demonstrates low-nanomolar activity against PLK1 (IC50 ~9 nM)
  • Supplied as a solid powder for preparing stock solutions
  • Available in milligram pack sizes, including 50 mg
  • High purity suitable for research applications (99.6%)
Catalog No. 50-002-31894 Supplier Medchemexpress LLC Supplier No. HY1203750MG
Rigosertib sodium is a research reagent and multi-kinase inhibitor used in preclinical studies to investigate cell cycle regulation, DNA damage response, and apoptosis. It is provided as a solid powder for preparation of stock solutions and is supplied in milligram-scale packs for laboratory use.
  • Multi-kinase inhibitor targeting PLK and PI3K pathways
  • Demonstrates low-nanomolar activity against PLK1 (IC50 ~9 nM)
  • Supplied as a solid powder for preparing stock solutions
  • Available in milligram pack sizes, including 50 mg
  • High purity suitable for research applications (99.6%)