Learn More
Enzo Life Sciences [pSer95, Ala97] RII (81-99) (0.5 mg)
SDP
Supplier:  Enzo Life Sciences BMLP1600500
Encompass_Preferred
Calcineurin substrate. This sequence, DLDVPIPGRFDRRVpSVAAE is derived from cAMP-dependent protein kinase regulatory subunit type II [PKA; [Ala-97]-RII (81-99)] with a phosphoserine at Ser95. This peptide can be used directly as a calcineurin (protein phosphatase 2B, PP2B) substrate. Phosphate release can be quantified using the (BML-AK111) detection reagent. Alternative Name: RII Phosphopeptide, [pSer95, Ala97] cAMP dependent protein kinase regulatory subunit type II (81 99). MW: 2192.0. Formulation: Lyophilized. Trifluoroacetate salt. Purity: ≥95% (HPLC). Sequence Asp-Leu-Asp-Val-Pro-Ile-Pro-Gly-Arg-Phe-Asp-Arg-Arg-Val-pSer-Val-Ala-Ala-Glu. Appearance: Solid. Solubility: Soluble in water. Long Term Storage: -20°C. Use/Stability: Stock solution in water is highly stable at -20°C. Handling: Protect from moisture
Catalog No. NC1035643