Learn More
Medchemexpress LLC HY-102004 10mg Medchemexpress, Rilapladib CAS:412950-08-4 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HY10200410MG
Encompass_Preferred
Medchemexpress, HY-102004 10mg Rilapladib CAS:412950-08-4 Rilapladib (SB 659032) is a selective Lipoprotein-Associated Phospholipase A2 (Lp-PLA2) inhibitor with an IC50 of 230 pM. Rilapladib (SB 659032) is also a PAFR (Platelet Activating Factor Receptor) antagonist. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-196-7947