Learn More
Medchemexpress LLC Rilmazafone hydrochloride | 85815-37-8 | 511.79 | 100 MG
SDP
Catalog No. 5000446756
Encompass_Preferred
Rilmazafone hydrochloride is a benzodiazepine (omega) ligand, intended for research use only.
  • Solid storage at 4°C
  • Sealed storage, away from moisture
  • In solvent storage at -80°C for 6 months
  • In solvent storage at -20°C for 1 month
Catalog No. 50-004-46756 Supplier Medchemexpress LLC Supplier No. HYU00228100MG
Rilmazafone hydrochloride is a benzodiazepine (omega) ligand, intended for research use only.
  • Solid storage at 4°C
  • Sealed storage, away from moisture
  • In solvent storage at -80°C for 6 months
  • In solvent storage at -20°C for 1 month