Learn More
Medchemexpress LLC Rilmazafone hydrochloride | 85815-37-8 | 511.79 | 25 MG
SDP
Catalog No. 5000449893
Encompass_Preferred
Rilmazafone hydrochloride is a benzodiazepine (omega) ligand. This product is for research use only and not sold to patients. It appears as a white to off-white solid and is soluble in DMSO at 250 mg/mL.
  • Acts as a benzodiazepine (omega) ligand.
  • For research use only.
  • Appears as a white to off-white solid.
  • Soluble in DMSO at 250 mg/mL.
  • Store solid form at 4°C in sealed storage, away from moisture.
  • Store in solvent at -80°C for 6 months or -20°C for 1 month in sealed storage, away from moisture.
Catalog No. 50-004-49893 Supplier Medchemexpress LLC Supplier No. 25MGHYU00228
Rilmazafone hydrochloride is a benzodiazepine (omega) ligand. This product is for research use only and not sold to patients. It appears as a white to off-white solid and is soluble in DMSO at 250 mg/mL.
  • Acts as a benzodiazepine (omega) ligand.
  • For research use only.
  • Appears as a white to off-white solid.
  • Soluble in DMSO at 250 mg/mL.
  • Store solid form at 4°C in sealed storage, away from moisture.
  • Store in solvent at -80°C for 6 months or -20°C for 1 month in sealed storage, away from moisture.