Learn More
Medchemexpress LLC Rilmazafone hydrochloride | 85815-37-8 | MFCD31544396 | 511.79 | 10 MM * 1 ML
SDP
Catalog No. 5000448468
Encompass_Preferred
Rilmazafone hydrochloride is a benzodiazepine (omega) ligand. It is intended for research use only and stimulates hepatic drug-metabolizing enzymes in various animal models, including rats, mice, and dogs.
  • Benzodiazepine (omega) ligand
  • Stimulates hepatic drug-metabolizing enzymes
  • For research use only
  • White to off-white solid appearance
  • Soluble in DMSO
Catalog No. 50-004-48468 Supplier Medchemexpress LLC Supplier No. HYU0022810MM/1ML
Rilmazafone hydrochloride is a benzodiazepine (omega) ligand. It is intended for research use only and stimulates hepatic drug-metabolizing enzymes in various animal models, including rats, mice, and dogs.
  • Benzodiazepine (omega) ligand
  • Stimulates hepatic drug-metabolizing enzymes
  • For research use only
  • White to off-white solid appearance
  • Soluble in DMSO