Learn More
Selleck Chemical LLC Rilmenidine Phosphate
SDP
Catalog No. 5000004157
Encompass_Preferred
Rilmenidine Phosphate is a selective I(1) imidazoline receptor agonist used for the treatment of hypertension
Catalog No. 50-000-04157 Supplier Selleck Chemical LLC Supplier No. S75485MG
Rilmenidine Phosphate is a selective I(1) imidazoline receptor agonist used for the treatment of hypertension