Learn More
TARGETMOL CHEMICALS INC RILMENIDINE PHOSPHATE 10MG
SDP
Catalog No. 502576482
Encompass_Preferred
Also available in 1 mg 5 mg 25 mg 50 mg 100 mg 500 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. Rilmenidine Phosphate is a selective I(1) imidazoline receptor agonist used to treat hypertension. purity: 99%
Catalog No. 50-257-6482 Supplier TARGETMOL CHEMICALS INC Supplier No. T664110MG
Also available in 1 mg 5 mg 25 mg 50 mg 100 mg 500 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. Rilmenidine Phosphate is a selective I(1) imidazoline receptor agonist used to treat hypertension. purity: 99%